3-(9-Carbazolyl)benzeneboronic acid, 98%
3-(9-Carbazolyl)benzeneboronic acid, 98%
3-(9-Carbazolyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

3-(9-Carbazolyl)benzeneboronic acid, 98%

CAS: 864377-33-3 | C18H14BNO2 | 287.125 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64898.03
also known as H64898-03
Price (USD)
54.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.30
Each
Chemical Identifiers
CAS864377-33-3
IUPAC Name[3-(9H-carbazol-9-yl)phenyl]boronic acid
Molecular FormulaC18H14BNO2
InChI KeyIDQUIFLAFFZYEX-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5% (HPLC)
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only