4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%
4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%
4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

4-Fluoro-2-nitro-5-(trifluoromethyl)aniline, 98%

CAS: 179062-05-6 | C7H4F4N2O2 | 224.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64928.03
also known as H64928-03
Price (USD)
77.65
Online exclusive
86.00
Save 8.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Online exclusive
86.00
Save 8.35
Each
Chemical Identifiers
CAS179062-05-6
IUPAC Name4-fluoro-2-nitro-5-(trifluoromethyl)aniline
Molecular FormulaC7H4F4N2O2
InChI KeyWOSGMZUIHAFPSS-UHFFFAOYSA-N
SMILESNC1=C(C=C(F)C(=C1)C(F)(F)F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only