3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%
3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%
3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%
3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%
Thermo Scientific Chemicals

3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%

CAS: 72587-18-9 | C6H4ClF3N2 | 196.557 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64945.03
also known as H64945-03
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.60
Save 11.95 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.60
Save 11.95 (15%)
Each
Add to cart
Chemical Identifiers
CAS72587-18-9
IUPAC Name2-chloro-5-(trifluoromethyl)pyridin-3-amine
Molecular FormulaC6H4ClF3N2
InChI KeyHABKYPVMMHBOIW-UHFFFAOYSA-N
SMILESNC1=C(Cl)N=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It's employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only