2-Chloropyrimidine-5-boronic acid, 96%
2-Chloropyrimidine-5-boronic acid, 96%
2-Chloropyrimidine-5-boronic acid, 96%
Thermo Scientific Chemicals

2-Chloropyrimidine-5-boronic acid, 96%

CAS: 1003845-06-4 | C4H4BClN2O2 | 158.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64949.03
also known as H64949-03
Price (USD)
64.65
Online Exclusive
72.20
Save 7.55 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
72.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS1003845-06-4
IUPAC Name(2-chloropyrimidin-5-yl)boronic acid
Molecular FormulaC4H4BClN2O2
InChI KeyYTCIHPTZKKWKKC-UHFFFAOYSA-N
SMILESOB(O)C1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only