4-Bromoimidazole, 98%
4-Bromoimidazole, 98%
4-Bromoimidazole, 98%
Thermo Scientific Chemicals

4-Bromoimidazole, 98%

CAS: 2302-25-2 | C3H3BrN2 | 146.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64972.06
also known as H64972-06
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS2302-25-2
IUPAC Name5-bromo-1H-imidazole
Molecular FormulaC3H3BrN2
InChI KeyFHZALEJIENDROK-UHFFFAOYSA-N
SMILESBrC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. C. H. Chen; B. A. Donatelli. Dihydro-1,4-dithiin annelation. J. Org. Chem.1976 41(18), 3053-3054.
  2. Karl-Erland Stensio; Kerstin Wahlberg and Robert Wahren. Synthesis of brominated imidazoles. Acta chemica scandinavica.1973 272179-2183.