(±)-3-Hydroxypyrrolidine hydrochloride, 98%
(±)-3-Hydroxypyrrolidine hydrochloride, 98%
(±)-3-Hydroxypyrrolidine hydrochloride, 98%
(±)-3-Hydroxypyrrolidine hydrochloride, 98%
Thermo Scientific Chemicals

(±)-3-Hydroxypyrrolidine hydrochloride, 98%

CAS: 86070-82-8 | C4H10ClNO | 123.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64975.14
also known as H64975-14
Price (USD)
170.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
170.00
Each
Add to cart
Chemical Identifiers
CAS86070-82-8
IUPAC Namepyrrolidin-3-ol hydrochloride
Molecular FormulaC4H10ClNO
InChI KeyQPMSJEFZULFYTB-UHFFFAOYNA-N
SMILESCl.OC1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only