L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals
L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals
L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals
L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H65046.06
also known as H65046-06
5 g
Catalog number H65046.06
also known as H65046-06
Price (USD)
683.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
683.00
Each
Add to cart
Chemical Identifiers
CAS3303-34-2
IUPAC Name(2S)-2-[(2S)-2-aminopropanamido]-4-methylpentanoic acid
Molecular FormulaC9H18N2O3
InChI KeyRDIKFPRVLJLMER-WZTWBHKBNA-N
SMILESCC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only