4-(Methylamino)benzonitrile, 97%
4-(Methylamino)benzonitrile, 97%
4-(Methylamino)benzonitrile, 97%
Thermo Scientific Chemicals

4-(Methylamino)benzonitrile, 97%

CAS: 4714-62-9 | C8H8N2 | 132.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H65148.06
also known as H65148-06
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS4714-62-9
IUPAC Name4-(methylamino)benzonitrile
Molecular FormulaC8H8N2
InChI KeyJEDXDYXIRZOEKG-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)benzonitrile is used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Krystyna Rotkiewicz; Wolfgang Rettig; Norbert Detzer and Annette Rothe. Substituent-induced coupling of the two lowest excited singlet states of 2-methoxy-derivatives of 4-(N,N-dimethylamino)- and 4-(N-methylamino)benzonitrile. Phys. Chem. Chem. Phys.2003, 5, 998-1002.