3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid
3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid
3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid
Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid

CAS: 109-54-6 | C5H12ClN | 121.608 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H65371.06
also known as H65371-06
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS109-54-6
IUPAC Namehydrogen (3-chloropropyl)dimethylamine chloride
Molecular FormulaC5H13Cl2N
InChI KeyLJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid or low melting solid
Assay (GC)≥96%
Identification (FTIR)Conforms
GC/MS AnalysisConforms
View more
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Roderic P. Quirk; Kwansoo Han; and Youngjoon Lee. Anionic synthesis of ω-dimethylamino-functionalized polymers by functionalization of polymeric organolithiums with 3-dimethylaminopropyl chloride. Polymer International.1999 48(2), 99-108.
  2. A. Keith Davies; Suppiah Navaratnam; and Glyn O. Phillips.Photochemistry of chlorpromazine [2-chloro-N-(3-dimethylaminopropyl)phenothiazine] in propan-2-ol solution. J. Chem. Soc. Perkin Trans. 2.1976 2(1), 25-29.