Ethyl 2-(bromomethyl)acrylate, 97%
Ethyl 2-(bromomethyl)acrylate, 97%
Ethyl 2-(bromomethyl)acrylate, 97%
Ethyl 2-(bromomethyl)acrylate, 97%
Thermo Scientific Chemicals

Ethyl 2-(bromomethyl)acrylate, 97%

CAS: 17435-72-2 | C6H9BrO2 | 193.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H65384.06
also known as H65384-06
Price (USD)
308.65
Online Exclusive
343.00
Save 34.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
308.65
Online Exclusive
343.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS17435-72-2
IUPAC Nameethyl 2-(bromomethyl)prop-2-enoate
Molecular FormulaC6H9BrO2
InChI KeyMTCMFVTVXAOHNQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=C)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless to pale yellow
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only