tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%
tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%
tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%
Thermo Scientific Chemicals

tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%

CAS: 35418-16-7 | C9H15NO3 | 185.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H65385.14
also known as H65385-14
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS35418-16-7
IUPAC Nametert-butyl 5-oxopyrrolidine-2-carboxylate
Molecular FormulaC9H15NO3
InChI KeyQXGSPAGZWRTTOT-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)C1CCC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥94.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only