2-(Methylamino)pyrimidine, 95%
2-(Methylamino)pyrimidine, 95%
2-(Methylamino)pyrimidine, 95%
Thermo Scientific Chemicals

2-(Methylamino)pyrimidine, 95%

CAS: 931-61-3 | C5H7N3 | 109.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H65564.03
also known as H65564-03
Price (USD)
191.65
Special offer
Online exclusive
Ends: 30-Jun-2026
225.00
Save 33.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
191.65
Special offer
Online exclusive
Ends: 30-Jun-2026
225.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS931-61-3
IUPAC NameN-methylpyrimidin-2-amine
Molecular FormulaC5H7N3
InChI KeyBQNXHDSGGRTFNX-UHFFFAOYSA-N
SMILESCNC1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only