2-(1-Imidazolyl)acetonitrile, 95%
2-(1-Imidazolyl)acetonitrile, 95%
2-(1-Imidazolyl)acetonitrile, 95%
2-(1-Imidazolyl)acetonitrile, 95%
Thermo Scientific Chemicals

2-(1-Imidazolyl)acetonitrile, 95%

CAS: 98873-55-3 | C5H5N3 | 107.116 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66012.03
also known as H66012-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS98873-55-3
IUPAC Name2-(1H-imidazol-1-yl)acetonitrile
Molecular FormulaC5H5N3
InChI KeyZPGCVVBPGQJSPX-UHFFFAOYSA-N
SMILESN#CCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.50%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only