(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%
(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%
(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%
Thermo Scientific Chemicals

(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%

CAS: 204512-95-8 | C4H10ClNO | 123.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66034.03
also known as H66034-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS204512-95-8
IUPAC Nameoxolan-3-amine hydrochloride
Molecular FormulaC4H10ClNO
InChI KeyMHOVLDXJDIEEMJ-UHFFFAOYNA-N
SMILESCl.NC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥94.0%
FormCrystalline powder or powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only