3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%

CAS: 832114-00-8 | C11H18BNO3 | 223.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66042.06
also known as H66042-06
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS832114-00-8
IUPAC Name3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Molecular FormulaC11H18BNO3
InChI KeyCVLHETBAROWASE-UHFFFAOYSA-N
SMILESCC1=C(B2OC(C)(C)C(C)(C)O2)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.8%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only