1-Ethyl-1-methylpyrrolidinium bromide, 98%
1-Ethyl-1-methylpyrrolidinium bromide, 98%
1-Ethyl-1-methylpyrrolidinium bromide, 98%
Thermo Scientific Chemicals

1-Ethyl-1-methylpyrrolidinium bromide, 98%

CAS: 69227-51-6 | C7H16BrN | 194.116 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H66050.22
also known as H66050-22
Price (USD)
233.65
Special offer
Online exclusive
Ends: 30-Jun-2026
275.00
Save 41.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
233.65
Special offer
Online exclusive
Ends: 30-Jun-2026
275.00
Save 41.35 (15%)
Each
Chemical Identifiers
CAS69227-51-6
IUPAC Name1-ethyl-1-methylpyrrolidin-1-ium bromide
Molecular FormulaC7H16BrN
InChI KeyKHJQQUGSPDBDRM-UHFFFAOYSA-M
SMILES[Br-].CC[N+]1(C)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.5%
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Pamela M. Dean; Bronya R. Clare; Vanessa Armel; Jennifer M. Pringle; Craig M. Forsyth; Maria Forsyth; Douglas R. MacFarlane. Structural Characterization of Novel Ionic Salts Incorporating Trihalide Anions. Australian Journal of Chemistry.2011, 62, (4), 334-340.