(5-Fluoro-2-methyl-3-indolyl)acetic acid, 97%, Thermo Scientific Chemicals
(5-Fluoro-2-methyl-3-indolyl)acetic acid, 97%, Thermo Scientific Chemicals
(5-Fluoro-2-methyl-3-indolyl)acetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(5-Fluoro-2-methyl-3-indolyl)acetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66088.03
also known as H66088-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein399422
CAS71987-67-2
Chemical Name or Material(5-Fluoro-2-methyl-3-indolyl)acetic acid
Formula Weight207.2
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only