Sodium 1-propanethiolate, tech. 85%
Sodium 1-propanethiolate, tech. 85%
Sodium 1-propanethiolate, tech. 85%
Sodium 1-propanethiolate, tech. 85%
Thermo Scientific Chemicals

Sodium 1-propanethiolate, tech. 85%

CAS: 6898-84-6 | C3H7NaS | 98.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66135.14
also known as H66135-14
Price (USD)
322.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
322.00
Each
Add to cart
Chemical Identifiers
CAS6898-84-6
IUPAC Namesodium propylsulfanide
Molecular FormulaC3H7NaS
InChI KeyXVSFHIIADLZQJP-UHFFFAOYSA-M
SMILES[Na+].CCC[S-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)95%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only