4-(Fmoc-amino)butyric acid, 95%
4-(Fmoc-amino)butyric acid, 95%
4-(Fmoc-amino)butyric acid, 95%
Thermo Scientific Chemicals

4-(Fmoc-amino)butyric acid, 95%

CAS: 116821-47-7 | C19H19NO4 | 325.364 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66147.06
also known as H66147-06
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.10
Save 12.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.10
Save 12.45
Each
Chemical Identifiers
CAS116821-47-7
IUPAC Name4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC19H19NO4
InChI KeyACUIFAAXWDLLTR-UHFFFAOYSA-N
SMILESOC(=O)CCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only