4-Chloroindole, 97%
4-Chloroindole, 97%
4-Chloroindole, 97%
Thermo Scientific Chemicals

4-Chloroindole, 97%

CAS: 25235-85-2 | C8H6ClN | 151.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66153.03
also known as H66153-03
Price (USD)
37.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.30
Each
Chemical Identifiers
CAS25235-85-2
IUPAC Name4-chloro-1H-indole
Molecular FormulaC8H6ClN
InChI KeySVLZRCRXNHITBY-UHFFFAOYSA-N
SMILESClC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid.
Assay from Supplier's CofA≥96.0%
Appearance (Color)Pale yellow to pale brown
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only