2-Amino-5-methylpyridine-3-carboxaldehyde, 95%
2-Amino-5-methylpyridine-3-carboxaldehyde, 95%
2-Amino-5-methylpyridine-3-carboxaldehyde, 95%
Thermo Scientific Chemicals

2-Amino-5-methylpyridine-3-carboxaldehyde, 95%

CAS: 1023814-35-8 | C7H8N2O | 136.154 g/mol
Have Questions?
Catalog NumberQuantity
H66175.MD
also known as H66175-MD
250 mg
Catalog number H66175.MD
also known as H66175-MD
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS1023814-35-8
IUPAC Name2-amino-5-methylpyridine-3-carbaldehyde
Molecular FormulaC7H8N2O
InChI KeyKHLBBDOMXLJWGR-UHFFFAOYSA-N
SMILESCC1=CN=C(N)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.90%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only