Chemical Identifiers
CAS32679-02-0
IUPAC Nameλ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene bis(acetonitrile) tetrafluoroboranuide
Molecular FormulaC12H18BF4N2Rh
InChI KeyKSMAFOANQHYEJG-SUESZSCISA-N
SMILES[Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1
View more