2-Oxetanemethanol, 96%
2-Oxetanemethanol, 96%
2-Oxetanemethanol, 96%
Thermo Scientific Chemicals

2-Oxetanemethanol, 96%

CAS: 61266-70-4 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66218.MD
also known as H66218-MD
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS61266-70-4
IUPAC Name(oxetan-2-yl)methanol
Molecular FormulaC4H8O2
InChI KeyPQZJTHGEFIQMCO-UHFFFAOYNA-N
SMILESOCC1CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid.
Assay (GC)≥95.0%
Proton NMRConforms
Appearance (Color)Colorless to pale yellow.
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
RUO – Research Use Only
P. Luger; J. Buschmann. Oxetane: the first x-ray analysis of a nonsubstituted four-membered ring. J. Am. Chem. Soc.1984106(23), 7118-7121.