2-(2-Cyanophenoxy)acetamide, 98%
2-(2-Cyanophenoxy)acetamide, 98%
2-(2-Cyanophenoxy)acetamide, 98%
Thermo Scientific Chemicals

2-(2-Cyanophenoxy)acetamide, 98%

CAS: 54802-12-9 | C9H8N2O2 | 176.175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H66271.14
also known as H66271-14
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
270.65
Online Exclusive
300.00
Save 29.35 (10%)
Each
Specifications
Beilstein2939357
CAS54802-12-9
Chemical Name or Material2-(2-Cyanophenoxy)acetamide
DOT InformationTransport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight176.17
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only