3-Amino-2-methoxypyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
3-Amino-2-methoxypyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
3-Amino-2-methoxypyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-2-methoxypyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66289.03
also known as H66289-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein19162520
CAS870997-81-2
Chemical Name or Material3-Amino-2-methoxypyridine-4-carboxylic acid
Formula Weight168.15
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only