3,5-Dimethyl-1-adamantaneacetic acid, 97%
3,5-Dimethyl-1-adamantaneacetic acid, 97%
3,5-Dimethyl-1-adamantaneacetic acid, 97%
3,5-Dimethyl-1-adamantaneacetic acid, 97%
Thermo Scientific Chemicals

3,5-Dimethyl-1-adamantaneacetic acid, 97%

CAS: 14202-14-3 | C14H22O2 | 222.328 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66357.03
also known as H66357-03
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.60
Save 12.95 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.60
Save 12.95 (15%)
Each
Add to cart
Specifications
CAS14202-14-3
Chemical Name or Material3,5-Dimethyl-1-adamantaneacetic acid
ColorWhite
Formula Weight222.32
IUPAC Name2-(3,5-dimethyl-1-adamantyl)acetic acid
View more
A commonly used as an organic reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only