(±)-2,3-Butanediol, 98%
(±)-2,3-Butanediol, 98%
(±)-2,3-Butanediol, 98%
Thermo Scientific Chemicals

(±)-2,3-Butanediol, 98%

CAS: 513-85-9 | C4H10O2 | 90.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number H66369.22
also known as H66369-22
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Chemical Identifiers
CAS513-85-9
IUPAC Name(2R,3S)-butane-2,3-diol
Molecular FormulaC4H10O2
InChI KeyOWBTYPJTUOEWEK-ZXZARUISSA-N
SMILESC[C@@H](O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4370-1.4420 @ 20°C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only