3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals
3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals
3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals
3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66400.MD
also known as H66400-MD
Price (USD)
77.65
Online Exclusive
86.60
Save 8.95 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.60
Save 8.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS209328-72-3
IUPAC Name5-chloro-6-fluoropyridin-3-ol
Molecular FormulaC5H3ClFNO
InChI KeyIETLNKVWDZYCPN-UHFFFAOYSA-N
SMILESOC1=CC(Cl)=C(F)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.10%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only