2-Bromo-1-naphthol, 95%
2-Bromo-1-naphthol, 95%
2-Bromo-1-naphthol, 95%
Thermo Scientific Chemicals

2-Bromo-1-naphthol, 95%

CAS: 771-15-3 | C10H7BrO | 223.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66429.03
also known as H66429-03
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.30
Save 13.65 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.30
Save 13.65 (14%)
Each
Chemical Identifiers
CAS771-15-3
SpecificationsSpecification SheetSpecification Sheet
FormLow melting powder
Appearance (Color)Off-white
Assay by NMR>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only