2-Amino-2'-chloro-5-nitrobenzophenone, 98%
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
Thermo Scientific Chemicals

2-Amino-2'-chloro-5-nitrobenzophenone, 98%

CAS: 2011-66-7 | C13H9ClN2O3 | 276.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H66440.14
also known as H66440-14
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS2011-66-7
IUPAC Name2-(2-chlorobenzoyl)-4-nitroaniline
Molecular FormulaC13H9ClN2O3
InChI KeyGRDGBWVSVMLKBV-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only