Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H66446.03
also known as H66446-03
1 g
Catalog number H66446.03
also known as H66446-03
Price (USD)
236.65
Online Exclusive
263.00
Save 26.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
236.65
Online Exclusive
263.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS156648-40-7
IUPAC Name(2S)-2-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC31H36N2O6
InChI KeyCNDRLQTWKWIHEP-KSYWNVGFNA-N
SMILESCC(N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to off-white
Assay (HPLC)>97.5%
Water Content (Karl Fischer Titration)<1%
Loss on Drying<1% (30?C, 2h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only