5-Aminophthalide, 97%, Thermo Scientific Chemicals
5-Aminophthalide, 97%, Thermo Scientific Chemicals
5-Aminophthalide, 97%, Thermo Scientific Chemicals
5-Aminophthalide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminophthalide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66457.03
also known as H66457-03
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS65399-05-5
IUPAC Name5-amino-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H7NO2
InChI KeyISMUWQMUWFPFBZ-UHFFFAOYSA-N
SMILESNC1=CC=C2C(=O)OCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormPowder or crystalline powder or solid
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only