Ethyl 6-chloroindole-2-carboxylate, 97%
Ethyl 6-chloroindole-2-carboxylate, 97%
Ethyl 6-chloroindole-2-carboxylate, 97%
Ethyl 6-chloroindole-2-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 6-chloroindole-2-carboxylate, 97%

CAS: 27034-51-1 | C11H10ClNO2 | 223.656 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66483.06
also known as H66483-06
Price (USD)
188.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
188.00
Each
Add to cart
Chemical Identifiers
CAS27034-51-1
IUPAC Nameethyl 6-chloro-1H-indole-2-carboxylate
Molecular FormulaC11H10ClNO2
InChI KeyFSMZLIBWSAMADK-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC=C(Cl)C=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Proton NMR≥96.0% - Conforms
Appearance (Color)Brown
Melting Point168.0-173.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only