Methyl 6-methoxyindole-2-carboxylate, 97%
Methyl 6-methoxyindole-2-carboxylate, 97%
Methyl 6-methoxyindole-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 6-methoxyindole-2-carboxylate, 97%

CAS: 98081-83-5 | C11H11NO3 | 205.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66528.03
also known as H66528-03
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS98081-83-5
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms
Appearance (Color)Pale yellow
FormFused solid
Assay from Supplier's CofA≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only