6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals
6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals
6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals
6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66548.03
also known as H66548-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein115231
CAS99071-54-2
Chemical Name or Material6-(Aminomethyl)quinoline
Formula Weight158.2
Health Hazard 1H302-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only