N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals
N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals
N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals
N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H66571.MD
also known as H66571-MD
250 mg
Catalog number H66571.MD
also known as H66571-MD
Price (USD)
33.70
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
33.70
Each
Add to cart
Chemical Identifiers
CAS82689-19-8
IUPAC Nametert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Molecular FormulaC16H22N2O3
InChI KeyJEFQUFUAEKORKL-LBPRGKRZSA-N
SMILESCC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only