Methyl indole-2-carboxylate, 97%
Methyl indole-2-carboxylate, 97%
Methyl indole-2-carboxylate, 97%
Methyl indole-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl indole-2-carboxylate, 97%

CAS: 1202-04-6 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66609.06
also known as H66609-06
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1202-04-6
IUPAC Namemethyl 1H-indole-2-carboxylate
Molecular FormulaC10H9NO2
InChI KeyNQPIEWBAWBFGOB-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormFused solid
Assay from Supplier's CofA≥96.0%
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only