2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
Thermo Scientific Chemicals

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%

CAS: 104206-65-7 | C14H10F3NO5 | 329.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H66633.MD
also known as H66633-MD
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS104206-65-7
IUPAC Name2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
Molecular FormulaC14H10F3NO5
InChI KeyOUBCNLGXQFSTLU-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1C(=O)C1C(=O)CCCC1=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.90%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only