1,2,3,4-Tetrahydro-2-naphthylamine, 97%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydro-2-naphthylamine, 97%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydro-2-naphthylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,4-Tetrahydro-2-naphthylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66652.06
also known as H66652-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Beilstein1102357
CAS2954-50-9
Chemical Name or Material1,2,3,4-Tetrahydro-2-naphthylamine
EINECS Number220-973-7
Formula Weight147.1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only