5-Amino-4-chloropyrimidine, 96%
5-Amino-4-chloropyrimidine, 96%
5-Amino-4-chloropyrimidine, 96%
5-Amino-4-chloropyrimidine, 96%
Thermo Scientific Chemicals

5-Amino-4-chloropyrimidine, 96%

CAS: 54660-78-5 | C4H4ClN3 | 129.547 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66671.03
also known as H66671-03
Price (USD)
117.65
Special offer
Online exclusive
Ends: 30-Jun-2026
138.00
Save 20.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
117.65
Special offer
Online exclusive
Ends: 30-Jun-2026
138.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS54660-78-5
IUPAC Name4-chloropyrimidin-5-amine
Molecular FormulaC4H4ClN3
InChI KeyLHGMCUVJFRBVBH-UHFFFAOYSA-N
SMILESNC1=CN=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
FormCrystals or crystalline powder or powder
Assay≥96.0% (GC)
Assay (unspecified)≥96.0% (GC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only