alpha-Bromo-4-fluorophenylacetic acid, 96%
alpha-Bromo-4-fluorophenylacetic acid, 96%
alpha-Bromo-4-fluorophenylacetic acid, 96%
Thermo Scientific Chemicals

alpha-Bromo-4-fluorophenylacetic acid, 96%

CAS: 29270-33-5 | C8H6BrFO2 | 233.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66692.03
also known as H66692-03
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.90
Save 9.25 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.90
Save 9.25 (15%)
Each
Specifications
CAS29270-33-5
Chemical Name or Materialalpha-Bromo-4-fluorophenylacetic acid
ColorWhite to Brown
DOT InformationTransport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight233.03
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only