5,6-Dihydroxyindole, 95%
5,6-Dihydroxyindole, 95%
5,6-Dihydroxyindole, 95%
Thermo Scientific Chemicals

5,6-Dihydroxyindole, 95%

CAS: 3131-52-0 | C8H7NO2 | 149.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66709.03
also known as H66709-03
Price (USD)
245.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
245.00
Each
Chemical Identifiers
CAS3131-52-0
IUPAC Name1H-indole-5,6-diol
Molecular FormulaC8H7NO2
InChI KeySGNZYJXNUURYCH-UHFFFAOYSA-N
SMILESOC1=C(O)C=C2C=CNC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to gray
FormSolid
Assay (unspecified)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only