(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%
(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%
(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%
Thermo Scientific Chemicals

(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%

CAS: 4187-56-8 | C8H10ClN | 155.625 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H66715.14
also known as H66715-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Beilstein2412316
Boiling Point232°C
CAS4187-56-8
Chemical Name or Material(S)-(-)-1-(4-Chlorophenyl)ethylamine
ColorYellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only