1-Benzhydryl-3-azetidinol, 97%
1-Benzhydryl-3-azetidinol, 97%
1-Benzhydryl-3-azetidinol, 97%
Thermo Scientific Chemicals

1-Benzhydryl-3-azetidinol, 97%

CAS: 18621-17-5 | C16H17NO | 239.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H66720.22
also known as H66720-22
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS18621-17-5
IUPAC Name1-(diphenylmethyl)azetidin-3-ol
Molecular FormulaC16H17NO
InChI KeyMMAJXKGUZYDTHV-UHFFFAOYSA-N
SMILESOC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.30%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only