Methyl indole-7-carboxylate, 97%
Methyl indole-7-carboxylate, 97%
Methyl indole-7-carboxylate, 97%
Thermo Scientific Chemicals

Methyl indole-7-carboxylate, 97%

CAS: 93247-78-0 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66729.03
also known as H66729-03
Price (USD)
37.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.00
Each
Chemical Identifiers
CAS93247-78-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream.
Assay (GC)≥96.0%
Proton NMRConforms
FormCrystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only