2-Amino-5-methoxypyrimidine, 97%
2-Amino-5-methoxypyrimidine, 97%
2-Amino-5-methoxypyrimidine, 97%
Thermo Scientific Chemicals

2-Amino-5-methoxypyrimidine, 97%

CAS: 13418-77-4 | C5H7N3O | 125.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66799.03
also known as H66799-03
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS13418-77-4
IUPAC Name5-methoxypyrimidin-2-amine
Molecular FormulaC5H7N3O
InChI KeyKAHHAPNRIQLSFT-UHFFFAOYSA-N
SMILESCOC1=CN=C(N)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Assay from Suppliers CofA≥96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only