5-Methyl-1-indanone, 97%
5-Methyl-1-indanone, 97%
5-Methyl-1-indanone, 97%
Thermo Scientific Chemicals

5-Methyl-1-indanone, 97%

CAS: 4593-38-8 | C10H10O | 146.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66814.14
also known as H66814-14
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Specifications
Beilstein119335
CAS4593-38-8
Chemical Name or Material5-Methyl-1-indanone
Formula Weight146.19
Health Hazard 1H302-H319
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only