2,5-Dimethylindole, 97%
2,5-Dimethylindole, 97%
2,5-Dimethylindole, 97%
Thermo Scientific Chemicals

2,5-Dimethylindole, 97%

CAS: 1196-79-8 | C10H11N | 145.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66874.06
also known as H66874-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1196-79-8
IUPAC Name2,5-dimethyl-1H-indole
Molecular FormulaC10H11N
InChI KeyZFLFWZRPMDXJCW-UHFFFAOYSA-N
SMILESCC1=CC2=CC(C)=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to dark brown
Assay (GC)≥96.0%
Proton NMRConforms
FormFused solid
Melting Point112-113?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only