Isoxazole-4-boronic acid pinacol ester, 97%
Isoxazole-4-boronic acid pinacol ester, 97%
Isoxazole-4-boronic acid pinacol ester, 97%
Isoxazole-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

Isoxazole-4-boronic acid pinacol ester, 97%

CAS: 928664-98-6 | C9H14BNO3 | 195.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66887.03
also known as H66887-03
Price (USD)
165.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
165.00
Each
Add to cart
Specifications
CAS928664-98-6
Chemical Name or MaterialIsoxazole-4-boronic acid pinacol ester
Formula Weight195.02
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only