N-Acetyl-L-tryptophan methyl ester, 95%
N-Acetyl-L-tryptophan methyl ester, 95%
N-Acetyl-L-tryptophan methyl ester, 95%
N-Acetyl-L-tryptophan methyl ester, 95%
Thermo Scientific Chemicals

N-Acetyl-L-tryptophan methyl ester, 95%

CAS: 2824-57-9 | C14H16N2O3 | 260.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66937.14
also known as H66937-14
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Add to cart
Specifications
CAS2824-57-9
Chemical Name or MaterialN-Acetyl-L-tryptophan methyl ester
ColorWhite
Formula Weight260.29
IUPAC Namemethyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only